Tenofovir disoproxil fumarate

Supplier: MORAVEK BIOCHEMICALS MS

Synonyms: (R)-(((((1-(6-Amino-9H-purin-9-yl)propan-2-yl)oxy)methyl)phosphoryl)bis(oxy))bis(methylene) diisopropyl dicarbonate fumarate

M1725-5MG M1725-1MG
101969-354EA 810.77 USD
101969-354 101969-352
Tenofovir disoproxil fumarate
Tenofovir disoproxil fumarate
Formula: C₁₉H₃₀N₅O₁₀P·C₄H₄O₄
MDL Number: MFCD08141829
CAS Number: 202138-50-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR