Saquinavir

Supplier: MORAVEK BIOCHEMICALS MS

Synonyms: (S)-N1-((2S,3R)-4-((3S,4aS,8aS)-3-(tert-butylcarbamoyl)octahydroisoquinolin-2(1H)-yl)-3-hydroxy-1-phenylbutan-2-yl)-2-(quinoline-2-carboxamido)succinamide, Ro-31-8959

M1616-1MG M1616-5MG M1616-10MG
101969-338EA 405.38 USD
101969-338 101969-340 101969-342
Saquinavir
Saquinavir
Formula: C₃₈H₅₀N₆O₅
MW: 670.84 g/mol
MDL Number: MFCD00866925
CAS Number: 127779-20-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR