Valacyclovir Hydrochloride Hydrate

Supplier: MORAVEK BIOCHEMICALS MS

Synonyms: L-Valine 2-(2-Amino-1,6-dihydro-6-oxo-9H-purin-9-ylmethoxy)ethyl Ester Hydrochloride

M1571-1MG M1571-5MG
101969-260EA 294.23 USD
101969-260 101969-262
Valacyclovir Hydrochloride Hydrate
Valacyclovir hydrochloride hydrate
Formula: C₁₃H₂₀N₆O₄
MW: 360.80 g/mol
MDL Number: MFCD01861507
CAS Number: 124832-27-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR