Ritonavir standard ≥98% (by HPLC), MORpure™ for stable isotope labelling (SIL), HPLC purified
Supplier: MORAVEK BIOCHEMICALS MS
Synonyms:
A-84538, Thiazol-5-ylmethyl ((2S,3S,5S)-3-hydroxy-5-((S)-2-(3-((2-isopropylthiazol-4-yl)methyl)-3-methylureido)-3-methylbutanamido)-1,6-diphenylhexan-2-yl)carbamate, ABT-538, Abbott 84538
M1535-1MG
M1535-5MG
101969-254EA
379.23
USD
101969-254
101969-256
Ritonavir standard ≥98% (by HPLC), MORpure™ for stable isotope labelling (SIL), HPLC purified
Ritonavir
Formula:
C₃₇H₄₈N₆O₅S₂ MW: 720.94 g/mol |
MDL Number:
MFCD00927142 CAS Number: 155213-67-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...