Tenofovir

Supplier: MORAVEK BIOCHEMICALS MS

M1502-1MG M1502-5MG
101969-246EA 353.08 USD
101969-246 101969-248
Tenofovir
Tenofovir
Formula: C₉H₁₄N₅O₄P
MDL Number: MFCD00943794
CAS Number: 147127-20-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR