Brilliant Green Agar Base with 1.2% Agar Dehydrated Culture Media, Granulated, HiMedia Laboratories
Supplier: HiMedia
GM016A-2.5KG GM016A-500G GM016A-5KG
75804-490EA 885.31 USD
75804-490 75804-492 75804-494
Brilliant Green Agar Base with 1.2% Agar Dehydrated Culture Media, Granulated, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Recommended as an enrichment medium for the isolation of Salmonellae from feces, urine, and other pathological materials.
  • Microbiological culture media
  • Packaged in 500 g poly bottles