Precision Tweezers with Ergonomic Handles, Apex Tool Group
Supplier: Cooper Tools
E2ASA E3CSA E5SA E7SA
37695-438EA 105.39 USD
37695-438 37695-440 37695-458 37695-442
Precision Tweezers with Ergonomic Handles, Apex Tool Group
Forceps
These tweezers are made of stainless steel, non-magnetic, non-rusting, acid-proof and heat-resistant with thin shaped tips to suit every application.
  • Thermally insulated, soft foam handles, ESD-safe
  • Ergonomically shaped handles reduce hand fatigue and facilitates comfortable working