cis-Dichlorobis(2,2'-bipyridine)ruthenium(II) dihydrate 99%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
cis-Bis(2,2'-bipyridine)dichlororuthenium(II) dihydrate
44-0200-5G
44-0200-1G
100206-632EA
628.43
USD
100206-632
100206-630
cis-Dichlorobis(2,2'-bipyridine)ruthenium(II) dihydrate 99%
cis-Dichlorobis(2,2'-bipyridine)ruthenium(II) dihydrate
Formula:
C₂₀H₂₀Cl₂N₄O₂Ru MW: 520.376 g/mol |
MDL Number:
MFCD00015504 CAS Number: 15746-57-3 |
Specification Test Results
Form | Crystalline |
Color | Black |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...