Albumin (Mouse) AlphaLISA Detection Kits, PerkinElmer
AL508F AL508C
101678-642EA 52461.54 USD
101678-642 101680-552
Albumin (Mouse) AlphaLISA Detection Kits, PerkinElmer
Assays Immunological Assays
The AlphaLISA® Mouse Albumin Detection Kit is designed for the quantitative detection of mouse albumin in serum, urine, buffered solution or cell culture medium, using a homogeneous (no wash steps, no separation steps) assay.
  • Harmonized Code US-3822005090
  • H phrases US-H317
  • P phrases US-P261-P280g-P272-P363-P333+P313-P302+P352a-P321-P501a
  • GHS-Signal words - US-Warning
  • GHS-Symbols - US-GHS07