PCSK9 (Human) AlphaLISA Detection Kits, PerkinElmer
101675-574EA 2382.6 USD
101675-574 101691-990 101679-218
PCSK9 (Human) AlphaLISA Detection Kits, PerkinElmer
Assays Immunological Assays
Formats: 500 assay point kit allows you to run 500 wells in 96-well or 384-well format, using a 50 µl reaction volume (5 µl of sample). 5000 assay point kit allows you to run 5000 wells in 96-well or 384-well format, using a 50 µl reaction volume (5 µl of sample). Features: no-wash steps, no separation steps, ELISA alternative technology, sensitive detection, broad sample compatibility, small sample volume, results in less than 3 hours, half the time of an ELISA assay. Human Proprotein Convertase Subtilisin / Kexin Type 9 (PCSK9) belongs to the proteinase K subfamily of the secretory subtilase family. This glycoprotein contains 692 amino acids, including a signal peptide, a prodomain, and a catalytic domain. Initially synthesized as a soluble 74 kDa precursor protein, it is cleaved into 14 kDa and 60 kDa domains, which remain associated. This protein plays a major regulatory role in cholesterol homeostasis and is highly expressed in the kidney, liver, and intestine. PCSK9 binds to the low-density lipoprotein receptor (LDLR), inducing LDLR degradation. Reduced LDLR levels result in decreased metabolism of low-density lipoprotein, which could lead to hypercholesterolemia. Inhibition of PCSK9 function is currently being explored as a means of lowering cholesterol levels. Mutations in this gene have been associated with a rare form of autosomal dominant familial hypercholesterolemia. AlphaLISA technology allows the detection of molecules of interest in a no-wash, highly sensitive, quantitative assay. In an AlphaLISA assay, a biotinylated anti-analyte antibody binds to the Streptavidin-coated donor beads while another anti-analyte antibody is conjugated to AlphaLISA acceptor beads. In the presence of the analyte, the beads come into close proximity. The excitation of the donor beads causes the release of singlet oxygen molecules that triggers a cascade of energy transfer in the acceptor beads, resulting in a sharp peak of light emission at 615 nm.
  • Harmonized Code US-3822005090
  • H phrases US-H302-H315-H318-H317-H402-H412
  • P phrases US-P261-P280a-P280i-P273-P264-P270-P272-P305+P351+P338-P310a-P301+P312a-P363-P332+P313-P333+P313-P337+P313-P330-P302+P352a-P321-P362+P364-P501a
  • GHS-Signal words - US-Danger
  • GHS-Symbols - US-GHS05 GHS07
Human Proprotein Convertase Subtilisin / Kexin Type 9 (PCSK9) standard, lyophilized, for use in running standard curves in AlphaLISA no-wash immunoassays. This standard is already provided in the AlphaLISA human PCSK9 kit, but can be ordered separately.