Insulin-like Growth Factor 2 (Human) AlphaLISA Detection Kits, PerkinElmer
101675-808EA 2382.6 USD
101675-808 101692-188 101679-548
Insulin-like Growth Factor 2 (Human) AlphaLISA Detection Kits, PerkinElmer
Assays Immunological Assays
Formats: 500 assay point kit allows you to run 500 wells in 96-well or 384-well format, using a 50 µl reaction volume (5 µl of sample). 5000 assay point kit allows you to run 5000 wells in 96-well or 384-well format, using a 50 µl reaction volume (5 µl of sample). Features: no-wash steps, no separation steps, ELISA alternative technology, sensitive detection, broad sample compatibility, small sample volume, results in less than 3 hours, half the time of an ELISA assay. Insulin-like Growth Factor 2 (IGF2) is 67 amino acids in length after being fully processed. Its actions are mediated by binding to specific cell surface receptors. The major role of IGF2 is as a growth promoting hormone during gestation. In general, the insulin-like growth factor pathway plays a major role in cancer cell proliferation, survival and resistance to anti-cancer therapies in many human malignancies. IGF2 could be potentially be an important serum biomarker for prognosis of certain cancers. AlphaLISA technology allows the detection of molecules of interest in a no-wash, highly sensitive, quantitative assay. In an AlphaLISA assay, a biotinylated anti-analyte antibody binds to the Streptavidin-coated donor beads while another anti-analyte antibody is conjugated to AlphaLISA acceptor beads. In the presence of the analyte, the beads come into close proximity. The excitation of the donor beads causes the release of singlet oxygen molecules that triggers a cascade of energy transfer in the acceptor beads, resulting in a sharp peak of light emission at 615 nm.
  • Harmonized Code US-3822005090
  • H phrases US-H302-H315-H318-H317-H402-H412
  • P phrases US-P261-P280a-P280i-P273-P264-P270-P272-P305+P351+P338-P310a-P301+P312a-P363-P332+P313-P333+P313-P337+P313-P330-P302+P352a-P321-P362+P364-P501a
  • GHS-Signal words - US-Danger
  • GHS-Symbols - US-GHS05 GHS07
Human Insulin-like Growth Factor 2 (IGF2) standard, lyophilized, for use in running standard curves in AlphaLISA no-wash immunoassays. This standard is already provided in the AlphaLISA human IGF2 kit, but can be ordered separately.