(R,R)-(-)-N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediaminocobalt(II), redish brown powder

Supplier: STREM CHEMICALS INC MS

Synonyms: (1R,2R)-(-)-1,2-Cyclohexanediamino-N,N'-bis(3,5-di-t-butylsalicylidene)cobalt (II), (R,R)-N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediaminocobalt(II), (R,R)-Jacobsen's catalyst cobalt(II) complex

27-0525-5G 27-0525-25G 27-0525-1G
100202-390EA 212.31 USD
100202-390 100196-930 100199-844
(R,R)-(-)-N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediaminocobalt(II), redish brown powder
(R,R)-(-)-N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediaminocobalt(II)

Jacobsen Ligand

Formula: C₃₆H₅₂CoN₂O₂
MW: 603.74 g/mol
Melting Pt: 406…412 °C
MDL Number: MFCD01631277
CAS Number: 176763-62-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR