Anti-PD-1 (CD279) Rat Monoclonal Antibody [clone: 29F.1A12]

Supplier: Bio X Cell
BP0273-25MG BP0273-50MG BP0273-5MG BP0273-100MG
103479-734EA 5100 USD
103479-734 103479-736 103479-738 103479-740 76554-522 76556-586 76557-272 76557-314
Anti-PD-1 (CD279) Rat Monoclonal Antibody [clone: 29F.1A12]
Antibodies
InVivoPlus anti-mouse PD-1 (CD279), Clone: 29F.1A12, Isotype: rat IgG2a, κ, Applications: in vivo blocking of PD-1/PD-L signaling, in vitro PD-1 neutralization, Immunohistochemistry (frozen), Flow cytometry, Western blot

Type: Primary
Antigen: PD-1 (CD279)
Clonality: Monoclonal
Clone: 29F.1A12
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a, kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR