Anti-TIGIT Mouse Monoclonal Antibody [clone: 1G9]

Supplier: Bio X Cell
BE0274-1MG BE0274-25MG BE0274-100MG BE0274-50MG BE0274-5MG
103254-368EA 3920 USD
103254-368 103254-370 103254-372 103254-374 103572-648 76555-478 76555-884 76556-868 76557-122 76557-220
Anti-TIGIT Mouse Monoclonal Antibody [clone: 1G9]
Antibodies
InVivoMab anti-mouse TIGIT, Clone: 1G9, Isotype: mouse IgG1, κ, Applications: in vivo TIGIT blockade, in vitro TIGIT blockade, Flow cytometry

Type: Primary
Antigen: TIGIT
Clonality: Monoclonal
Clone: 1G9
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR