Anti-IFNAR-1 Mouse Monoclonal Antibody [clone: MAR1-5A3]

Supplier: Bio X Cell
BP0241-50MG BP0241-5MG BP0241-25MG BP0241-100MG
103254-344EA 5100 USD
103254-344 103254-346 103254-348 103254-350 76555-308 76555-710 76556-082 76556-174
Anti-IFNAR-1 Mouse Monoclonal Antibody [clone: MAR1-5A3]
Antibodies
InVivoPlus anti-mouse IFNAR-1, Clone: MAR1-5A3, Isotype: mouse IgG1, κ, Applications: in vivo IFNAR-1 blockade, in vitro IFNAR-1 blockade

Type: Primary
Antigen: IFNAR-1
Clonality: Monoclonal
Clone: MAR1-5A3
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR