Anti-CD4 Rat Monoclonal Antibody [clone: GK1.5]

Supplier: Bio X Cell
BP0003-1-25MG BP0003-1-100MG BP0003-1-50MG BP0003-1-5MG
103254-328EA 5100 USD
103254-328 103254-330 103254-332 103254-334 76554-538 76555-528 76556-634 76556-688
Anti-CD4 Rat Monoclonal Antibody [clone: GK1.5]
Antibodies
InVivoPlus anti-mouse CD4, Clone: GK1.5, Isotype: rat IgG2b, κ, Applications: in vivo CD4+ T cell depletion, Flow cytometry

Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: GK1.5
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR