Anti-CD40L (CD154) Armenian Hamster Monoclonal Antibody [clone: MR-1]

Supplier: Bio X Cell
BP0017-1-25MG BP0017-1-5MG BP0017-1-50MG BP0017-1-100MG
103254-192EA 5100 USD
103254-192 103254-194 103254-196 103254-198 76555-424 76556-378 76556-776 76557-092
Anti-CD40L (CD154) Armenian Hamster Monoclonal Antibody [clone: MR-1]
Antibodies
InVivoPlus anti-mouse CD154 (CD40L), Clone: MR-1, Isotype: Armenian hamster IgG, Applications: in vivo blocking of CD40/CD40L signaling, in vitro blocking of CD40/CD40L signaling

Type: Primary
Antigen: CD40L (CD154)
Clonality: Monoclonal
Clone: MR-1
Conjugation:
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR