Anti-CXCR3 (CD183) Armenian Hamster Monoclonal Antibody [clone: CXCR3-173]

Supplier: Bio X Cell
BE0249-100MG BE0249-25MG BE0249-1MG BE0249-50MG BE0249-5MG
103254-136EA 3920 USD
103254-136 103254-138 103254-140 103254-142 103572-616 76554-548 76554-800 76555-202 76555-730 76556-088
Anti-CXCR3 (CD183) Armenian Hamster Monoclonal Antibody [clone: CXCR3-173]
Antibodies
InVivoMab anti-mouse CXCR3 (CD183), Clone: CXCR3-173, Isotype: Armenian hamster IgG, Applications: in vivo CXCR3 neutralization, Flow cytometry

Type: Primary
Antigen: CXCR3 (CD183)
Clonality: Monoclonal
Clone: CXCR3-173
Conjugation:
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR