Anti-Pan-endothelial Cell Antigen (MECA-32) Rat Monoclonal Antibody [clone: MECA-32]

Supplier: Bio X Cell
BE0200-25MG BE0200-50MG BE0200-5MG BE0200-1MG BE0200-100MG
103253-896EA 3920 USD
103253-896 103253-898 103253-900 103253-902 103572-906 76555-392 76556-588 76556-732 76556-738 76556-790
Anti-Pan-endothelial Cell Antigen (MECA-32) Rat Monoclonal Antibody [clone: MECA-32]
Antibodies
InVivoMab anti-mouse Pan-endothelial Cell Antigen (MECA-32), Clone: MECA-32, Isotype: rat IgG2a, κ, Applications: Immunohistochemistry (frozen), Flow cytometry, Immunofluorescence, Western blot

Type: Primary
Antigen: Pan-endothelial Cell Antigen (MECA-32)
Clonality: Monoclonal
Clone: MECA-32
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR