Anti-PD-L2 (B7-DC) Rat Monoclonal Antibody [clone: TY25]

Supplier: Bio X Cell
BE0112-1MG BE0112-5MG BE0112-50MG BE0112-25MG BE0112-100MG
103253-716EA 3920 USD
103253-716 103253-718 103253-720 103253-722 103572-530 76554-606 76555-262 76555-602 76555-706 76557-196
Anti-PD-L2 (B7-DC) Rat Monoclonal Antibody [clone: TY25]
Antibodies
InVivoMab anti-mouse PD-L2 (B7-DC), Clone: TY25, Isotype: rat IgG2a, κ, Applications: in vivo PD-L2 blockade, in vitro PD-L2 blockade, Immunohistochemistry (frozen), Flow cytometry

Type: Primary
Antigen: PD-L2 (B7-DC)
Clonality: Monoclonal
Clone: TY25
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR