Anti-CD86 (B7-2) Rat Monoclonal Antibody [clone: GL-1]

Supplier: Bio X Cell
BE0025-1MG BE0025-5MG BE0025-50MG BE0025-100MG BE0025-25MG
103253-430EA 3920 USD
103253-430 103253-432 103253-434 103253-436 103572-450 76554-418 76555-280 76555-404 76556-470 76556-812
Anti-CD86 (B7-2) Rat Monoclonal Antibody [clone: GL-1]
Antibodies
InVivoMab anti-mouse CD86 (B7-2), Clone: GL-1, Isotype: rat IgG2a, κ, Applications: In vivo CD86 blockade, Flow cytometry

Type: Primary
Antigen: CD86 (B7-2)
Clonality: Monoclonal
Clone: GL-1
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR