Anti-HA Mouse Monoclonal Antibody [clone: 12CA5]

Supplier: Bio X Cell
RT0268-1MG RT0268-25MG
103253-376EA 150 USD
103253-376 103253-378 103253-380 103253-382 76555-326 76556-064
Anti-HA Mouse Monoclonal Antibody [clone: 12CA5]
Antibodies
ReadyTag anti-HA, Clone: 12CA5, Isotype: mouse IgG2b, Applications: ELISA, Western blot, Immunoprecipitation, Immunofluorescence

Type: Primary
Antigen: HA
Clonality: Monoclonal
Clone: 12CA5
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR