Anti-PD-1 (CD279) Mouse Monoclonal Antibody [clone: J116]

Supplier: Bio X Cell
BE0188-5MG BE0188-25MG BE0188-50MG BE0188-1MG BE0188-100MG
103253-008EA 3920 USD
103253-008 103253-010 103253-012 103253-014 103572-886 76554-376 76555-064 76555-894 76556-258 76557-284
Anti-PD-1 (CD279) Mouse Monoclonal Antibody [clone: J116]
Antibodies
InVivoMab anti-human PD-1 (CD279), Clone: J116, Isotype: mouse IgG1, κ, Applications: in vitro PD-1 neutralization, in vivo PD-1 blockade in humanized mice

Type: Primary
Antigen: PD-1 (CD279)
Clonality: Monoclonal
Clone: J116
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR