Anti-v-H-Ras Rat Monoclonal Antibody [clone: Y13-238]

Supplier: Bio X Cell
BE0139-5MG BE0139-100MG BE0139-1MG BE0139-50MG BE0139-25MG
103252-756EA 3920 USD
103252-756 103252-758 103252-760 103252-762 103572-538 76555-170 76555-544 76556-500 76556-562 76557-102
Anti-v-H-Ras Rat Monoclonal Antibody [clone: Y13-238]
Antibodies
InVivoMab anti-mouse/human/rat v-H-Ras, Clone: Y13-238, Isotype: rat IgG2a, Applications: Immunoprecipitation

Type: Primary
Antigen: v-H-Ras
Clonality: Monoclonal
Clone: Y13-238
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse, Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR