Anti-MAdCAM-1 Rat Monoclonal Antibody [clone: MECA-367]

Supplier: Bio X Cell
BE0035-1MG BE0035-50MG BE0035-100MG BE0035-25MG BE0035-5MG
103252-454EA 3920 USD
103252-454 103252-456 103252-458 103252-460 103572-470 76555-122 76555-290 76555-624 76556-374 76556-918
Anti-MAdCAM-1 Rat Monoclonal Antibody [clone: MECA-367]
Antibodies
InVivoMab anti-mouse MAdCAM-1, Clone: MECA-367, Isotype: rat IgG2a, κ, Applications: in vivo MAdCAM-1 neutralization, Immunofluorescence

Type: Primary
Antigen: MAdCAM-1
Clonality: Monoclonal
Clone: MECA-367
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR