Anti-LFA-1alpha (CD11a) Rat Monoclonal Antibody [clone: FD441.8]

Supplier: Bio X Cell
BE0005-1-50MG BE0005-1-100MG BE0005-1-1MG BE0005-1-25MG BE0005-1-5MG
103252-214EA 3920 USD
103252-214 103252-216 103252-218 103252-220 103572-408 76554-504 76555-448 76555-666 76556-336 76557-180
Anti-LFA-1alpha (CD11a) Rat Monoclonal Antibody [clone: FD441.8]
Antibodies
InVivoMab anti-mouse LFA-1α (CD11a), Clone: FD441.8, Isotype: rat IgG2b, Applications: in vivo LFA-1 neutralization

Type: Primary
Antigen: LFA-1alpha (CD11a)
Clonality: Monoclonal
Clone: FD441.8
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR