Anti-CD40 Rat Monoclonal Antibody [clone: FGK4.5/FGK45]

Supplier: Bio X Cell
BE0016-2-1MG BE0016-2-100MG BE0016-2-50MG BE0016-2-5MG BE0016-2-25MG
103252-302EA 3920 USD
103252-302 103252-304 103252-306 103252-308 103572-430 76554-474 76555-142 76556-036 76556-544 76556-944
Anti-CD40 Rat Monoclonal Antibody [clone: FGK4.5/FGK45]
Antibodies
InVivoMab anti-mouse CD40, Clone: FGK4.5/FGK45, Isotype: rat IgG2a, Applications: in vivo CD40 activation, in vitro B cell stimulation/activation, in vitro DC stimulation/activation

  • Recommended Dilution Buffer: IP0065

Type: Primary
Antigen: CD40
Clonality: Monoclonal
Clone: FGK4.5/FGK45
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR