Anti-CD70 Rat Monoclonal Antibody [clone: FR70]

Supplier: Bio X Cell
BE0022-50MG BE0022-100MG BE0022-1MG BE0022-5MG BE0022-25MG
103252-358EA 3920 USD
103252-358 103252-360 103252-362 103252-364 103572-444 76554-672 76554-828 76554-972 76555-128 76555-574
Anti-CD70 Rat Monoclonal Antibody [clone: FR70]
Antibodies
InVivoMab anti-mouse CD70, Clone: FR70, Isotype: rat IgG2b, κ, Applications: In vivo CD70 blockade, In vitro CD70 blockade, Flow cytometry

Type: Primary
Antigen: CD70
Clonality: Monoclonal
Clone: FR70
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR