Lactobacillus MRS Agar Dehydrated Culture Media, HiMedia
Supplier: HiMedia
M641I-500G GM641-2.5KG GM641-500G GM641-5KG
95022-946EA 205.9 USD
95022-946 95022-948 89226-116 89226-118 75804-358 75804-360 75804-362
Lactobacillus MRS Agar Dehydrated Culture Media, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
Lactobacilli MRS Agar is recommended for the isolation, enumeration and cultivation of Lactobacillus spp.
  • Final pH ( at 25 °C) is 6.5 ±0.2
Certificaciones: The composition and performance criteria of M641I is according ISO 1995, Draft ISO/DIS 13720.