Fluid Thioglycollate Medium, Dehydrated, HiMedia Laboratories
Supplier: HiMedia
MAP009-500G M009-500G MAP009-500G MAP009-500G
95020-026EA 137.17 USD
95020-026 95020-024 95022-620 95022-682
Fluid Thioglycollate Medium, Dehydrated, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Dehydrated culture media for enrichment, isolation, cultivation, and maintenance of microorganisms.
  • Microbiological culture media
  • 500 g poly bottles