Luria Bertani Agar, Miller, Dehydrated Culture Media, HiMedia Laboratories
Supplier: HiMedia
M1151-2.5KG M1151-500G M1151-5KG M1151-10KG
89129-482EA 741.37 USD
89129-482 95020-584 89135-064 89223-128
Luria Bertani Agar, Miller, Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
For the cultivation and maintenance of recombinant strains of Escherichia coli for genetic and molecular studies.
  • Microbiological culture media
  • 500 g poly bottles
Suitable for routine isolation cultivation of microorganisms that are not particularly fastidious.