M17 Broth Dehydrated Culture Media, HiMedia Laboratories
Supplier: HiMedia
M1029-500G GM1029-2.5KG GM1029-500G GM1029-5KG
95020-326EA 318.5 USD
95020-326 75803-320 75803-322 75803-324
M17 Broth Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
For the cultivation and isolation of lactic streptococci and bacteriophages.
  • Microbiological culture media