Cooked Meat Medium (R.C. Medium) Dehydrated Culture Media, HiMedia
Supplier: HiMedia
M149-500G M149S-500G
95021-222EA 412.26 USD
95021-222 95021-232
Cooked Meat Medium (R.C. Medium) Dehydrated Culture Media, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
For the cultivation of aerobes and anaerobes, especially pathogenic Clostridia.
  • Microbiological culture media
  • 500 g poly bottles
Also for the maintenance of stock cultures.