Antimicrobial Inhibitor Test Agar Dehydrated Culture Media, HiMedia Laboratories
Supplier: HiMedia
M1601-500G
M1631-500G
M1632-500G
95021-324EA
308.16
USD
95021-324
95021-380
95021-382
Antimicrobial Inhibitor Test Agar Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology
Dehydrated Media for Microbiology
For the residual analysis of antimicrobial components in meat and organ samples using Bacillus subtilis (ATCC 6633) and Micrococcus luteus (ATCC 9341) as test organisms
- Microbiological culture media
- Packaged in 500 g poly bottles