SHOWA S-TEX350 A4 Nitrile Palm Coated, Kevlar/Hagane Steel, Cut Resistant Glove, Showa
Supplier: Showa
S-TEX350S-07 S-TEX350M-08 S-TEX350L-09 S-TEX350XL-10
10852-326CS 2114.81 USD
10852-326 10852-328 10852-330 10852-332
SHOWA S-TEX350 A4 Nitrile Palm Coated, Kevlar/Hagane Steel, Cut Resistant Glove, Showa
Gloves
100% palm dipped Nitrile glove, with Kevlar/Hagane steel provides excellent protection from cuts, abrasions, and punctures.
  • A4 cut resistant
  • Nitrile palm dipped
These gloves are used when a dry grip and resistance to cuts are required. Ergonomic design provides extreme comfort and dexterity.
Certificaciones: ANSI Cut Level A4.