ZING Green Safety Eco Safety Sign, Caution X-Ray Area, English/Spanish
Supplier: ZING Enterprises
1930 1930A 1930S
76023-314EA 25.28 USD
76023-314 76023-316 76023-318
ZING Green Safety Eco Safety Sign, Caution X-Ray Area, English/Spanish
Signs
The sign is available in one size 10" x 7" and three eco-friendly materials.
  • UL Validated for recycled content
  • Made in USA
  • High quality
Plastic signs are made from a minimum of 70% post-consumer recycled materials and are printed with UV-resistant inks
Aluminum signs are made from 94% recycled materials. The signs are printed with UV-resistant inks, and are suitable for indoor and outdoor use. The signs are pre-drilled for easy mounting.
Adhesive signs are made from a minimum of 70% post-industrial polystyrene, and are resistant to solvents, moisture, scuffing, tears and UV exposure.
All Zing signs are made in the USA from recycled materials. Recycled materials support sustainability initiatives and LEED credits. The sign meets OSHA and ANSI standards. Signs are sold 1/pack.