Mucicarmine stock solution

Supplier: Electron Microscopy Sciences

26080-05 26080-06
100504-462EA 129.34 USD
100504-462 100504-464
Mucicarmine stock solution
Mucicarmine

Mucicarmine is for use with hematoxylin and metanil yellow for the determination of carbohydrates and mucosubstances.

MDL Number: MFCD00071289
CAS Number: 51395-97-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR