Tris(hydroxypropyl)phosphine ≥80%

Synonyms: THPP, THP, Tris(3-hydroxypropyl)phosphine

777854-1G 777854-5G
102281-926EA 56.97 USD
102281-926 102281-928
Tris(hydroxypropyl)phosphine ≥80%
Tris(hydroxypropyl)phosphine
Formula: C₉H₂₁O₃P
MW: 208.24 g/mol
MDL Number: MFCD00269835
CAS Number: 4706-17-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR