Tris(hydroxypropyl)phosphine ≥80%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
THPP, THP, Tris(3-hydroxypropyl)phosphine
15-6375-10G
15-6375-50G
100209-316EA
285.91
USD
100209-316
100209-318
Tris(hydroxypropyl)phosphine ≥80%
Tris(hydroxypropyl)phosphine
Phosphine, Phosphine, Water Soluble, Surfactants for Nano Synthesis
Formula:
C₉H₂₁O₃P MW: 208.24 g/mol |
MDL Number:
MFCD00269835 CAS Number: 4706-17-6 UN: 2922 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...