Hairnets, Nylon, Cellucap-Melco
Supplier: Cellucap-Melco
HN500DB HN500WT HN500LB
33500-302CS 468.73 USD
33500-302 33500-304 33500-308
Hairnets, Nylon, Cellucap-Melco
Disposable Garments
Hairnets are made from a single strand, 15 denier nylon with 1/4" aperture
  • Blends seamlessly with similar colored hair for a discreet look
  • Individually tissued to avoid tangling
The garment is also referred to as the "invisible net", since it blends seamlessly with similar colored hair for a discreet look.