Doxycycline hydrochloride
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms:
(4S,4aR,5S,5aR,6R,12aS)-4-(Dimethylamino)-3,5,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-1,4,4a,5,5a,6,11,12a-octahydrotetracene-2-carboxamide hydrochloride
Doxycycline is an antibiotic - against gram-positive and gram-negative bacteria. More active biologically than the corresponding 6b-epimer hydrochloride.
Doxycycline is an antibiotic - against gram-positive and gram-negative bacteria. More active biologically than the corresponding 6b-epimer hydrochloride.
Derivative of oxytetracycline. A broad spectrum inhibitor used to inhibit matrix metallo-proteinases (MMP) such as type 1 collagenase in studies on would healing and tissue remodeling.
Doxycycline is a member of the tetracycline antibiotics group, and is commonly used to treat a variety of infections. It is a Broad spectrum antibiotic and bacteriostatic. Doxycycline shows antiprotozoal properties and is a potent inhibitor of MMPs (matrix metalloproteinases) in vivo. Doxycycline is frequently used to treat Lyme disease, chronic prostatitis, sinusitis, pelvic inflammatory disease acne, rosacea, and rickettsial infections and prophylaxis of anthraxDoxycycline (caused by Bacillus anthracis). It is also effective against Yersinia pestis. Treatment with doxycycline inhibits collagen synthesis.
Precaución: Storage Temperature: +4 °C, desiccate, protect from light
Formula:
C₂₂H₂₄N₂O₁₀·HCl MW: 480.9 g/mol |
MDL Number:
MFCD03427564 CAS Number: 10592-13-9 |
Specification Test Results
Physical Appearance | Yellow Powder |
pKa | (10 mg/ml) 2-3 |
Solubiity | Soluble in water (10 mg/mL - clear, yellowish to yellowish-green solution) and DMSO: soluble (50 mg/mL). |
Typical Working Concentration | It is typically used at a concentration of 20 ug/mL and is stable in media at 37 °C for approximately 5 days. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...