Cefoperazone sodium salt 870 - 1015 µg/mg, white powder USP
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms:
Cefoneg, Cefosint
Cefoperazone sodium salt is used to study drug-protein binding, expression and inhibition of penicillin-binding proteins (PBPs) during cell wall synthesis.
Cefoperazone sodium salt is a broad spectrum third generation cephalosporin antibiotic has similar antimicrobial activity to ceftazidime. It is active against Pseudomonoas aeruginosa and it has enhanced activity against Enterobacteriaceae and Bacteroides sp. in the presence of the beta-lactamase inhibitor sulbactam. Compared to cefotaxime, cefoperazone has greater susceptibility to hydrolysis by some beta-lactamases.
Cefoperazone exerts its bactericidal effect by inhibiting the bacterial cell wall synthesis. Cefoperazone has a dual excretory pattern, primarily via the biliary system and secondarily via the kidney
Formula:
C₂₅H₂₆N₉NaO₈S₂ MW: 667.648 g/mol |
MDL Number:
MFCD07793331 CAS Number: 62893-20-3 |
Specification Test Results
Physical Appearance | White powder |
Solubility | Soluble in water (50 mg/mL - clear, faint yellow solution). |
pH | (1 in 4) 4.5 - 6.5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...