Doxycycline hyclate 800 - 920 µg/mg USP
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms:
Doxycycline Hydrochloride Hemiethanolate Hemihydrate
It is used to inhibit matrix metalloproteinases and has been used to inhibit the inflammatory response of Lyme Disease.
Doxycycline is a relative of tetracycline reported to be more effective against sensitive organisms.
Derivative of oxytetracycline. A broad spectrum inhibitor used to inhibit matrix metallo-proteinases (MMP) such as type 1 collagenase in studies on would healing and tissue remodeling. Doxycycline hyclate inhibits the inflammatory response to the Lyme Disease Spirochete Borrelia burgdorferi. It is a broad spectrum inhibitor of matrix metalloproteinases in vivo.
Formula:
C₂₂H₂₄N₂O₈·HCl·0.5H₂O·0.5C₂H₆O MW: 512.94 g/mol (503.94 anhy) |
MDL Number:
MFCD07357237 CAS Number: 24390-14-5 |
|
Specification Test Results
Physical Appearance | Bright Yellow Powder |
pH | 2.0 - 3.0 (10 mg/mL water) |
Water Content | 1.4 - 2.8% |
Solubility | 50 mg/mL in water. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...