Earle’s balanced salt solutions (EBSS) Formula II
Supplier: Electron Microscopy Sciences
11766-10 11766-05
100496-418EA 49.33 USD
100496-418 100496-416
Earle’s balanced salt solutions (EBSS) Formula II
Buffers Cell Culture Buffers
All of our Earle’s Balanced Salt Solution contains: Potassium Chloride, Sodium Chloride, Sodium Phosphate Monobasic, and D-glucose. Some formulations may contain additional components. All of the solutions are prepared 1X strength.
Earle’s Balanced Salt Solutions Formula II contains:
KCl: 0.152 M
NaCl: 0.152 M
NaH₂PO₄·H₂O: 0.152 M
MgSO₄·7H₂O: 0.152 M
CaCl₂,·2H₂O: 0.152 M
NaHCO₃: 0.152 M
D-Glusose: 0.152 M
Phenol Red: 0.015 g/L