Thymidine (2'-deoxythymidine) ≥99% cell culture reagent

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: 1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidine-2,4-dione, 1-(2-Deoxy-β-D-ribofuranosyl)thymine, 1-(2-Deoxy-β-D-ribofuranosyl)-5-methyluracil, dT, Thymine deoxyriboside, 1-((2R,4S,5R)-4-Hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-methylpyrimidine-2,4(1H,3H)-dione, Pyrimidine deoxynucleoside, Thymidine kinase, Thymidine-2-deoxythymidine

0219475401 0219475405 0219475410 0219475425
IC0219475401EA 71.33 USD
IC0219475401 IC0219475405 IC0219475410 IC0219475425
Thymidine (2'-deoxythymidine) ≥99% cell culture reagent
Thymidine (2'-deoxythymidine)

Thymidine is a nucleoside composed of deoxyribose (a pentose sugar) joined to the pyrimidine base thymine.Deoxythymidine can be phosphorylated with one, two or three phosphoric acid groups, creating respectively dTMP, dTDP or dTTP (deoxythymidine mono- di- or triphosphate.It exists in solid form as small white crystals or white crystalline powder.The stability of deoxythymidine under standard temperature and pressure (STP) is very high.Thymidine is listed as a chemical teratogen.
Mainly used in forming hybrid cell lines wherein cells are incubated in HAT medium(hypoxanthine-aminopterin-thymidine medium) to give rise to monoclonal antibodies. Use in HT and HAT cocktails for hybridoma fusion, selection and cloning.
Physical Appearance: White Powder
Optical Rotation: (c=1, water) +17.5 - +19.5°
UV/Visible Absorbance: λmax (0.1N HCl) 266 - 268 nm
Solubility: Soluble in water, methanol, hot alcohol, hot acetone, hot ethyl acetate pyridine, glacial acetic acid and HCl (20 mg/mL-clear, colorless solution); sparingly soluble in hot chloroform

Formula: C₁₀H₁₄N₂O₅
MW: 242.23 g/mol
Melting Pt: 185…191 °C
Storage Temperature: Ambient
MDL Number: MFCD00006537
CAS Number: 50-89-5
EINECS: 200-070-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=124385dc87f82ac9484dc155601df1ef729452fd&vid=d4c7b9b5721827337436af7e49bc10f8fdad1953&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR