Dextran, M.W. 60,000-90,000 clinical grade

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

0218014010 0218014050 0218014080 0218014090 0218014091
IC0218014010EA 77.66 USD
IC0218014010 IC0218014050 IC0218014080 IC0218014090 IC0218014091
Dextran, M.W. 60,000-90,000 clinical grade
Dextran

Use of dextrans as long and hydrophilic spacer arms improves the performance of immobilized proteins acting on macromolecules. Dextrans are used in many applications as platelet aggregants, plasma volume extenders, osmotic pressure regulators, stabilizers, organ separation media, matrix components, copolymers, microcarriers, binding agents, viscosity modifiers, antithrombotics, lubricants and physical structure components. They may be used as long hydrophilic spacer arms to improve the performance (freedom of movement) of conjugated/bound proteins. Dextrans may be derivatized for use in biosensor systems.
Dextran is branched polysaccharide made up of linear α (1→6) linked glucose units and α (1→3) link initiated branches; it ranges in size from 10,000 to 150,000 Kd.Depending upon its molecular weight it has different uses in different fields.
Physical Appearance: White powder
Solubility: Very soluble in water (50 mg/mL), slightly soluble in alcohol

Formula: (C₆H₁₀O₅)n
Storage Temperature: Ambient
MDL Number: MFCD00130935
CAS Number: 9004-54-0
EINECS: 232-677-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR