Calcium chloride dihydrate 99.0-107.0% USP
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
02199524.5
0219952405
IC02199524.5EA
117.37
USD
IC02199524.5
IC0219952405
Calcium chloride dihydrate 99.0-107.0% USP
Calcium chloride dihydrate
Calcium chloride is useful in preparing competent bacteria. Calcium chloride dihydrate was used in the preparation of calcium-alginate beads. It was used to compose the medium for Thermophilic Clostridia (MTC). It may be used for the concentration of virus vectors in a protocol.
Store at Room Temperature (15-30 °C). Store Desiccated.
Physical Appearance: White Powder
Solubility: Soluble in water with heat liberation, ethanol, acetic acid, acetone.
pH: 4.5-9.2 (5% aq soln)
Heavy Metals: ≤0.001%
Formula:
CaCl₂·2H₂O Melting Pt: 176 °C (dec., lit.) Storage Temperature: Ambient |
MDL Number:
MFCD00149613 CAS Number: 10035-04-8 EINECS: 233-140-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...