2'-Deoxy-5-methylcytidine

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

Synonyms: 4-Amino-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-methylpyrimidin-2(1H)-one, 5-mdC, 5-Methyl-2'-deoxycytidine, 1-(2-Deoxy-beta-D-ribofuranosyl)-5-methylcytosine

0219888310 0219888380
IC0219888310EA 74.39 USD
IC0219888310 IC0219888380
2'-Deoxy-5-methylcytidine
2'-Deoxy-5-methylcytidine

5-Methyl-2'-deoxycytidine in single-stranded DNA can act in cis to signal de novo DNA methylation.
Physical Appearance: White Powder
Vapor Pressure: 8.85E-12mmHg at 25 °C (lit.)
Optical Rotation: [α]25/D +63°, c = 1 in 1 M NaOH (lit.)
Refractive Index: 1.694 (lit.)
Solubility: Soluble in 1 N NaOH

Formula: C₁₀H₁₅N₃O₄
MW: 241.25 g/mol
Melting Pt: 217…219 °C (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00006549
CAS Number: 838-07-3
EINECS: 212-655-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8c3c0fb735bcc9ac8eeb8cacc8084edada448a14&vid=8c3c0fb735bcc9ac8eeb8cacc8084edada448a14&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR