Sodium perchlorate monohydrate NaClO485.0-90.0 ACS
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
|
Danger
|
Synonyms:
Perchloric acid sodium salt
0219486280
0219486290
IC0219486280EA
79.92
USD
IC0219486280
IC0219486290
Sodium perchlorate monohydrate NaClO485.0-90.0 ACS
Sodium perchlorate monohydrate
Sodium Perchlorate Monohydrate is used in standard DNA extraction and hybridization reactions in molecular biology.
Physical Appearance: White Crystalline Powder
pH: 6.0 - 8.0 (5% aq soln)
Solubility: Very soluble in water
Formula:
NaClO₄·H₂O MW: 140.46 g/mol Melting Pt: 130 Density: 2.02 g/mL (lit.) Storage Temperature: Ambient |
MDL Number:
MFCD00149164 CAS Number: 7791-07-3 UN: 1502 ADR: 5.1 II Merck Index: 12,08798 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...