2-Oleoyl-1-palmitoyl-Sn-glycero-3-phospho-rac-(1-glycerol)sodium salt ≥99%

Supplier: AVANTI POLAR LIPIDS INC MS

Synonyms: POPG, 1-Palmitoyl-2-oleyl-sn-glycero-3-phospho-rac-(1-glycerol) sodium salt, POPG sodium salt, 1-Palmitoyl-2-oleoyl-sn-glycero-3-phospho-(1'-rac-glycerol) (sodium salt), POPG-Na, 1-Palmitoyl-2-oleoyl phosphatidylglycerol sodium salt, Sodium 2,3-dihydroxypropyl ((R)-2-(oleoyloxy)-3-(palmitoyloxy)propyl) phosphate(Z), Sodium 2,3-Dihydroxypropyl [2-(Oleoyloxy)-3-(palmitoyloxy)propyl] phosphate

840457P500MG 840457P25MG 840457P200MG 840457P1G
100127-250EA 807.69 USD
100127-250 100124-932 100120-794 100125-918
2-Oleoyl-1-palmitoyl-Sn-glycero-3-phospho-rac-(1-glycerol)sodium salt ≥99%
2-Oleoyl-1-palmitoyl-Sn-glycero-3-phospho-rac-(1-glycerol)sodium salt

Synonyms: POPG; PG(16:0/18:1(9Z)); 1-hexadecanoyl-2-(9Z-octadecenoyl)-sn-glycero-3-phospho-(1'-rac-glycerol) (sodium salt).


  • Stability: 6 Months
  • Storage -20°C

Formula: C₄₀H₇₆NaO₁₀P
MW: 771.002 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00674290
CAS Number: 268550-95-4

Order Now

Specification Test Results

Purity 99%
Percent composition C 62.31%, H 9.94%, Na 2.98%, O 20.75%, P 4.02%
Transition temperature -2 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR